Radial Purge Extension Window, PerkinElmer
Catalog #: CA97012-920
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Radial purge extension window for Optima
- Cat. no.:CA97012-920
- Supplier No.:N0770322
Specifications
About this item
This quartz radial window for the purge extension is used with Optima™ 4x00 DV and pre- November 2004 5x00 DV ICP-OES series instruments.
The Radial View Purge Extension Window is a replaceable quartz window that slides into the mount between the torch and the optics for the radial view.
Customers Who Bought This Also Bought