Distillation Rods, Büchi
Catalog #: 76169-032
Supplier: Buchi
Specifications
- Description:Distillation Rods, Set of 10
Specifications
About this item
Reusable glass rods designed specifically to aid in digestion of samples for Kjeldahl determination.
- Prevent bumping
- Minimize foaming
- Designed not to interfere with aspiration of automated systems
Aids in minimizing foaming of high surface tension samples to keep entire sample contents within the reflux zone. Helps to equilibrate heat evenly throughout the samples.
Customers Who Bought This Also Bought