Reversible Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
The super durable reversible microcentrifuge tube racks accommodate 0.2, 0.5, 1.5 and 2.0 ml microcentrifuge tubes.
- Autoclavable polypropylene
- Excellent rack for benchtop use or freezer storage
- 96 well racks have a translucent cover to allow viewing of samples while they are protected.
The 80/60 well rack capacities: 80-place for 1.5 and 2.0 ml tubes, 60-place for 0.2 and 0.5 ml tubes.
The 96 well rack capacities: 96-place for 1.5 and 2.0 ml tubes, 96-place for 0.2 and 0.5 ml tubes.
Customers Who Bought This Also Bought