Kjeldahl Weighing Boats, N-free, Buchi
Catalog #: 76169-030
Supplier: Buchi
Specifications
- Dimensions:58×10×10 mm
- Description:Kjeldahl Weighing Boats, 100 Pcs.
- Cat. no.:76169-030
Specifications
About this item
Weighing boats for powder, solids or viscous fluids for reliable nitrogen and protein determination according to Kjeldahl.
- Convenient handling and easy transfer into sample tube
- Reliable results thanks to high quality
- Nitrogen-free paper
Simply tare the boat, weigh and record sample, place boat into tube and raise tube to slide boat to the bottom of the vessel. Add acid and Kjeldahl tablet catalyst.
Customers Who Bought This Also Bought