About this item
For the quantification of progesterone in plasma and other sample matrices
- Measure progesterone in plasma and other sample matrices
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure progesterone levels down to 10 pg/ml
- Rapid assay; get results in under 4 hours
- Midpoint: 50% B/B0: 40-90 pg/ml
Progesterone, along with pregnenolone, is the biosynthetic precursor of all other steroid hormones. The main function of progesterone is to prepare the uterine lining for implantation of a fertilized ovum and to maintain pregnancy. Measurement of serum or plasma progesterone levels are used as an index to monitor ovulation and investigate luteal function.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:
- Environmentally Preferable:
Specifications
Related Information
AT-582601.pdf
Datasheet
Cayman Chemical Steroid Hormone Assay.pdf