Vydac 218MS C18, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules.
- Five reversed-phase chemistries
- Excellent peak shape with little or no TFA
- High protein recoveries make scale-up easy
- USP L1
Polymeric bonding, highest hydrophobic interaction and unique geometric selectivity. Use for simple enzymatic digests (<12 proteins) or biomolecules 0 to 5k MW.
A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- 75812-972
- 75812-974
- 75812-976
- 75831-416
- 75799-092
- 75831-420
- 75812-970
- 75831-408
- 75831-412
- 75831-404
- 75798-078
- 75798-076
- 75799-082
- 75799-078
- 75799-090
- 75799-086
- 75799-088
- 75830-784
- 75799-074
- 75799-076
- 75831-424
- 75799-072
- 75798-054
- 75798-060
- 75798-058
- 75798-064
- 75798-062
- 75799-060
- 75799-062
- 75798-066
- 75799-058
- 75799-068
- 75798-068
- 75799-070
- 75798-074
- 75798-072
- 75831-204
- 75831-200
- 75798-038
- 75799-094
- 75799-096
- 75798-042
- 75799-106
- 75799-102
- 75799-104
- 75798-044
- 75831-400
- 75798-052
- 75798-050
- Carbon load:
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1