Duran Connection Cap, GL 45, 3 Ports, Wheaton, DWK Life Sciences
Catalog #: 75995-256
Supplier: DWK Life Sciences
Specifications
- Description:Duran Cap with Steel 3 Ports
- Cat. no.:75995-256
- Thread Size:GL 45
Specifications
About this item
A robust and durable tubing connection system for use with all sizes of DURAN® GL 45 laboratory and media bottles
Ideal for chemistry, life science and biopharma laboratories.
The bottle top adaptor facilitates the transfer of liquids within a closed and sterile system.
Customers Who Bought This Also Bought