XPR Excellence Precision Balances, Mettler Toledo
Supplier: Mettler Toledo
Certificates
About this item
XPR Precision Balances, successor to the XPE Precision Balances, deliver outstanding weighing performance and support the highest requirements for data integrity and compliance. Quality assurance features ensure your weighing results are always valid, helping you to comply with even the strictest regulations.
- 7" color capacitive touchscreen terminal, glove compatible
- Internal calibration
- SmartPan weigh pan technology on 0.01, 0.001, and 0.0001 g balances
- Storage of weighing methods (including sample series and tolerances)
- Integrated results notepad
- GWP Approved built-in quality assurance monitoring
- Customizable tolerance profiles
- MinWeigh warning function
- Adjustment and routine test history
- Balance ready StatusLight™
- LevelControl: level warning with graphical leveling guide
- Temperature and time programmable automatic adjustment (proFACT)
- User Management, individual profiles password protected
- Change history log
- Easy connectivity and data export: 4×USB, 1 LAN
- Easy connection of peripheral devices: printers, PC, barcode readers, footswitches and sensors
- Integrated label editor, alphanumeric and barcode
- Export of process and results data to XML or Excel
- Integrated overload protection
- Chemically resistant materials
- Designed for easy cleaning
LabX automatically saves all results and process information securely in a central database.
The extraordinary stability provided by the new Monobloc™ weighing cell as well as the rugged balance design means you get your results fast even under challenging production environments.
Certifications: This ensures traceability and assists with FDA 21 CFR part 11 compliance. UL listed and CSA certified.
Specifications
- Catalog No:
- 76097-988
- 76097-944
- 76097-990
- 76097-946
- 76097-984
- 76097-980
- 76097-982
- 76097-974
- 76097-970
- 76097-972
- 76097-968
- 76098-006
- 76097-962
- 76098-008
- 76097-964
- 76097-958
- 76098-004
- 76097-960
- 76097-998
- 76098-000
- 76097-956
- 76097-996
- 76097-952
- 76097-954
- 76097-992
- 76097-948
- 76097-994
- 76097-950
- Communication interface:
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
- 4 x USB, LAN, RS232 Compatible
Specifications
Related Information
Mettler Toledo Dinohunt Mettler-Toledo Balance or Analytical Instrument
Promotion Material
Customers Who Bought This Also Bought