ContactUsLinkComponent
PHCbi Cell IQ™ Dry Heat Sterilization CO₂ Incubator, PHC Corporation
PHCbi Cell IQ™ Dry Heat Sterilization CO₂ Incubator, PHC Corporation
Certificates
PHCbi Cell IQ™ Dry Heat Sterilization CO₂ Incubator, PHC Corporation
PHCbi Cell IQ™ Dry Heat Sterilization CO₂ Incubator, PHC Corporation
Certificates
Specifications
  • Catalog No:
  • CA75856-512
  • Recovery time after 30 second door opening at 37 °C:
  • 15 min
  • CO₂ recovery time after 30 second door opening at 5 Vol.% CO₂:
  • 5 min
  • Energy consumption:
  • 376
  • Temperature range:
  • Ambient +5…50 °C
  • No. of shelves:
  • 4 (stainless steel copper enriched alloy) supplied as standard (Max. 10); 470W×450D×12H mm, Max. load 7 kg/shelf (15 lbs.)
  • Electrical:
  • 115V, 60Hz
  • Display:
  • Color LCD touchscreen readable to 0.1 increments