About this item
Long Ranger™ sequencing reagents are designed for the high performance sequencing on ABI Prism® 377 and Li-COR® Sequencers.
- Long Ranger™ gel solution offers expceptional results for sequencing and STR analysis
- Applications: DNA and RNA sequencing, STR analysing, identify testing, genotyping, different display
- Storage temperature upto 18 - 26 °C
Single™ pack contains all necessary reagent for casting a single gel in a pre-measures, ready to use pouch. Gel™ Slick solution is a safe alternative to hazardous silance based coating solution and it is suitable for sequencing gel and coating microcentrifuge tubes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.