Staticmaster® Electrostatic Discharge Workstation Kit with 2U500 Ionizer, NRD
Catalog #: 75794-920
Supplier: NRD Staticmaster
Specifications
- Description:ESD workstation kit with 2U500 ionizer
- Cat. no.:75794-920
- Supplier No.:SMPW-KITA
Specifications
About this item
This Staticmaster® PW basics kit is essential for eliminating static charge in the workspace and providing in-chamber ionization.
- Eliminates static charge
- Provides in-chamber ionization
Includes the ESD workstation kit, (1) 2U500 ionizer, (1) mount, and (1) 5060 grounding kit.
Caution: Eliminates static charge
Provides in-chamber ionization
Customers Who Bought This Also Bought