Staticmaster® Unistat Positioner, NRD
Catalog #: 58582-003
Supplier: NRD Staticmaster
Specifications
- Description:Staticmaster Unistat Positioner
- Cat. no.:58582-003
- Supplier No.:BF1
Specifications
About this item
The Staticmaster® BF1-500 Unistat Positioner quickly and effectively removes static charges from powders, filters, and other thin membrane materials that need to be precisely weighed.
- Quick and effective
- Convenient flexible neck
- Useful for powders, filters, and other thin membrane materials
The positioner's flexible neck makes this device convenient to use in any laboratory setting. The BF1-500 includes a ground wire attachment to connect to the ground tab of a single 2U500 Ionizing Cartridge and includes the model 5060 Ground Kit. By adding the ground kit, the performance of the positioner is greatly enhanced.
Caution: Quick and effective
Convenient flexible neck
Useful for powders, filters, and other thin membrane materials
Customers Who Bought This Also Bought