Staticmaster® I-Beam Stand Ionizer and Ionizer Kit, NRD
Supplier: NRD Staticmaster
Certificates
About this item
The Staticmaster® 6105 I-Beam Stand Ionizer Kit is a "shockless" electrical static eliminator ideally suited for removing static charge from fine powders, filter media, and irregularly shaped items prior to being placed in the weigh chamber
- "Shockless" electrical static eliminator
- Useful for filter media, powders, and irregularly shaped objects
- Removes static charges outside of weigh chamber
The I-Beam Stand Ionizer is a vertically mounted bar ionizer that is utilized outside of the weigh chamber. The Kit comes complete with an I-Beam Stand Ionizer and 7.5 KV Power Supply.
Ordering information: This system is equipped with a 6 foot, shielded power supply cable and is capable of supporting a second I-Beam.
Caution: "Shockless" electrical static eliminator
Useful for filter media, powders, and irregularly shaped objects
Removes static charges outside of weigh chamber