Pierce™ UltraLink™ Hydrazide Resin Affinity Chromatography Resin, Thermo Scientific
Catalog #: CAPI53149
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:UltraLink™ Hydrazide Resin
- Size:10 mL
- Cat. no.:CAPI53149
- Supplier No.:53149
Specifications
About this item
UltraLink™ Hydrazide resin is used to prepare affinity columns with antibodies or other glycolsylated proteins.
- Able to immobilise 1 to 10 mg of oxidized antibody or other glycoprotein per mL of resin
- No stabilisation step is required
- Conjugates only to purified glycoproteins whose sugar groups have been gently oxidised with periodate
- Immobilise in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
Formulation: Hydrazide-activated UltraLink Resin, slurried in water with sodium azide
Customers Who Bought This Also Bought