SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Catalog #: CA95055-940L
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:>40 µm
- pH range:4 - 13
- Description:SP Sephadex™ C-25
- Size:100 g
- Matrix:Cross-linked dextran
- Ligand:Sulphopropyl
- Binding capacity:lgG (Mr 160 000)- 1.1 mg/ml drained medium), Bovine COHb (Mr 69 000)- 70 mg/ml drained medium), Ribonuclease (Mr 13 700)- 230 mg/ml drained medium)
- Total ionic capacity:2 - 2,6 mmol/g
- Cat. no.:CA95055-940L
- Type:Solid
- Supplier No.:17-0230-01
Specifications
About this item
SP Sephadex™ C-25 is a strong cation exchanger.
The ion exchange group is a sulphopropyl group which remains charged and maintains consistently high capacity over the entire working range of pH 4 to 13.
Customers Who Bought This Also Bought