Steelite™ FW57, All Weather Safety Boots, Lace-Up, Portwest
Supplier: Portwest
Certificates
About this item
This waterproof ankle boot features a waterproof and breathable membrane, nubuck leather upper and a PU/TPU outsole. It is perfect for use in all conditions.
- Protective steel toecap
- Puncture-resistant steel midsole
- Direct injected dual density construction
- Lightweight EVA cushioned midsole
- Slip-, fuel-, and oil-resistant and heat resistant TPU/TPU outsole
- Waterproof membrane
- Removable EVA cushion footbed
- Anti-static
Steel toecap and midsole for total protection with a tough TPU outsole for greater stability and abrasion.
Color: Black
Certifications: ASTM F2413-11
Customers Who Bought This Also Bought