Specifications
- Description:Bi-Directional Needle-less Injection Site with Male Luer Lock
- Cat. no.:89134-222
- Supplier No.:LLV
Specifications
About this item
SAI offers a variety of Luer Valves and Swabbable Injection Caps to simplify catheter access and prevent the introduction of microbial pathogens.
- Male Luer for easy catheter or harness connection
- All parts provided ETO-exposed
These caps can be used anywhere that catheter access is needed, such as harnesses, extension lines, catheters, and Huber needles.
Needle less injection site with male Luer lock are made of polycarbonate, silicone.