Superdex Columns, 30 Increase, Cytiva
Supplier: Cytiva
Certificates
About this item
Superdex™ 30 Increase columns provide high resolution separations under mild conditions for small biomolecules having a molecular weight between 100 and 7000, such as peptides and oligosaccharides.
- Runtime reduced down to one third with maintained resolution for faster results
- Up to 50% higher resolution for improved purity and analysis results
- Higher lot-to-lot consistency
Specifications
- Catalog No:
- CA76015-028
- CA76015-026
- Max. pressure:
- <3.0 MPa [30 bar] (435 psi)
- <3.0 MPa [30 bar] (435 psi)
- pH range:
- 3 - 12
- 3 - 12
- Storage:
- 4 to 30 °C, 20% ethanol
- 4 to 30 °C, 20% ethanol
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought