HiScreen MabSelect Xtra Columns
Catalog #: CA97067-792
Supplier: Cytiva
Specifications
- Capacity:4.7 ml
- Max. pressure:3 bar [0.3 MPa] (44 psi)
- Particle size:75 µm
- Description:HiScreen™ MabSelect Xtra™
- pH range:3 - 10
- Binding capacity:Approx. 40 mg humanlgG/mL at 2.4 min residence time
- Storage:2...8 °C, 20% Ethanol
- Column Hardware:PP
- Bed Dimensions:7.7×100 mm
- Cat. no.:CA97067-792
- Supplier No.:28926976
Specifications
About this item
HiScreen™ MabSelect Xtra™ is prepacked with MabSelect Xtra™ of 4.7 ml volume and bed height of 10 cm make HiScreen™ columns excellent tools for method optimisation, parameter screening for capture of monoclonal antibodies (MAbs), robustness testing, and convenient scale-up.
- Easily connected in series to achieve 20 cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Flow velocity <300 cm/h; The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used
- Dynamic capacity: ~ 40 mg human IgG/ml medium
Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.
Customers Who Bought This Also Bought