Sodium dihydrogen phosphate, anhydrous 98.0-102%, Hi-AR™
Catalog #: CA10662-018
Supplier: HiMedia
CAS Number: 7558-80-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:500 g
- Cat. no.:CA10662-018
- Supplier No.:GRM3964-500G
Specifications
Specification Test Results
- Appearance:White to off-white crystals or powder or chunks
- Solubility:100 mg soluble in 1 mL of water
- pH (1M in water at 25°C):4.00 - 6.00
- Chloride (Cl):≤ 0.014%
- Sulfate (SO₄):≤ 0.15%
- Loss on drying (at 60°C for 1h, then 105°C for 4 h):≤ 2.00%
- Assay (NaOH Titration):98.00 - 102.00%
Specification Test Results
About this item
Synonyms: Sodium dihydrogenphosphate (granular),Sodium dihydrogen orthophosphate,Monobasic Sodium Phosphate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: ~ 650 °C
:Flash Pt:
:Flash Pt:
:M.W. 119.98 g/mol
:Sodium dihydrogen phosphate
Formula:NaH₂PO₄
MW:119.98 g/mol
MDL Number:MFCD00003527
EINECS:231-449-2
:650
Storage Temperature:Ambient
:12,08806
CAS Number:7558-80-7
Customers Who Bought This Also Bought