SW® PowerForm® PF-95LG Light Green 5.9 mil Nitrile Exam EnerGel® Rejuvenating Gloves
Supplier: SW Safety Solutions
Certificates
About this item
Work with confidence in premium gloves that combine tactile sensitivity with trusted protection, in addition to EnerGel™ technology which soothes and hydrates skin.
- EnerGel® hand conditioning technology hydrates and soothes skin; proven to prevent many skin ailments
- EcoTek® sustainable technology
- Breach Alert® visual detection technology provides durable 2-ply construction and contrasting inner color
- Proven resistance to fentanyl and gastric acid for over 4 hours to replicate hazardous settings and overdose conditions
- Fully textured for secure grip
- Silicone-free for critical environments
- 1.5 AQL exceeds laboratory standards
Work with confidence in premium gloves that combine tactile sensitivity with trusted protection, plus EnerGel® technology to soothe and hydrate skin. ASTM tested against viral penetration with low dermatitis potential. EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; Meets stringent standards that ensure skin safety; EAC TP TC 019/2011; CE 0493; CE EN 455.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).
Specifications
- Catalog No:
- 76053-592
- 76053-088
- 76053-092
- Model:
- PF-95LG
- PF-95LG
- PF-95LG
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Food contact approved:
- Yes
- Yes
- Yes
Specifications
Customers Who Bought This Also Bought