Specifications
- Description:5 Way InstaTest® Drinking Water Test Strips
- Cat. no.:470228-504
- Measurement Range:Nitrate 0-200 ppm, Nitrite 0-10 ppm, pH 6-9, Alkalinity 0-240 ppm, and Hardness 0-180 ppm
- No. of Tests:25 Strips/Vial
Specifications
About this item
Keep it simple with these easy-to-use, individual water test strips.
- Easy to use
- No additional reagents needed
A great way to monitor a variety of common water quality factors without having to measure reagents. Compare your readings to a handy color comparator chart printed right on the storage vial.