Friction Block Sand Paper
Catalog #: 470230-072
Supplier: Wards
Specifications
- Description:Friction Block Sand Paper
- Cat. no.:470230-072
Specifications
About this item
Friction blocks are useful for teaching the basics of force and motion.
- Wooden friction block with c-hook attached at one end
- Use with a spring scale to measure force and mass - all the numbers you need to solve a variety of motion and force problems
- Differing pine and sandpaper surfaces allow for comparative study
- Hands-on, economic tool for teaching fundamental physics concepts to students of any age
- Manufactured in the US
Friction blocks are useful for teaching the basics of force and motion in a simple, yet effective way. Fitted with a c-hook, the block can be attached to a spring scale to measure incident forces. Slide the block along a flat surface, or pull it up an incline. Observe the block by itself, or add weights to the top. Measure differences in coefficient of friction between the sandpaper and pine surfaces. A friction block used in tandem with a spring scale is a powerful tool for teaching students about friction, kinetic and potential energy, acceleration, and force. This block is sourced and manufactured in the United States.
Customers Who Bought This Also Bought