Order Entry
ContactUsLinkComponent
Puritan® Wooden Skewer, Pointed Tip, Puritan Medical Products
Puritan® Wooden Skewer, Pointed Tip, Puritan Medical Products
Catalog #: 75993-414
Applicator Wooden Stick, white birch, Non-sterile, 8in x 1/8in sold in Bulk
Puritan® Wooden Skewer, Pointed Tip, Puritan Medical Products
Catalog #: 75993-414
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Length:
    8" (203.2 mm)
  • Description:
    Wooden Skewer, Pointed Tip, Non Sterile
  • Cat. no.:
    75993-414

Specifications

About this item

Wooden skewers are made for general purpose functionality

  • High-quality applicator
  • General-purpose use
  • Made in USA

Choose a size and shape that works best for required applications, such as specimen transfer, mixing rods, and laboratory work. Sticks are manufactured from Northern Maine White Birch

Customers Who Bought This Also Bought