USB B/M to B/M Cable, Eisco Scientific
Catalog #: 470229-986
Supplier: Wards
Specifications
- Description:USB b/m to b/m
- Cat. no.:470229-986
Specifications
About this item
1m Neulog B/M to B/M Cable.
- USB 2.0 converter cable takes Type B Male connection to Type B Male connection
- 1 meter in length
USB 2.0 Cable - Type B Male to Type B Male - Converter Cable, 1m. Standard USB 2.0 B/M to B/M cable.
Customers Who Bought This Also Bought