Order Entry
ContactUsLinkComponent
LB Agar Plates with 25 µg/ml Kanamycin, X-gal and IPTG, Teknova
LB Agar Plates with 25 µg/ml Kanamycin, X-gal and IPTG, Teknova
Catalog #: CA100219-340
Supplier:  Teknova
CAS Number:  
LB Agar Plates with 25 µg/ml Kanamycin, X-gal and IPTG, Teknova
Catalog #: CA100219-340
Supplier:  Teknova
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    25 µl g/mL plus 60 µl g/mL X-Gal and 0.1 mM IPTG
  • Size:
    100 mm
  • Cat. no.:
    CA100219-340
  • Supplier No.:
    L1923

Specifications

About this item

For use with kanamycin resistant strains and strains harboring plasmids such as pPIC9K, pCR®-Blunt, pUC9P series plasmids. X-gal allows blue/white selection of colonies.

  • Sleeve of 20 plates
  • Sterile

Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 25 µg/ml kanamycin, 0.1 mM IPTG, 60 µg/ml X-gal.

Ambient shipping conditions. Refrigerate to store.

Customers Who Bought This Also Bought