To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:LB Agar, Lennox with 25 µg/mL Zeocin
- Size:100 mm
- Cat. no.:CA100219-984
- Supplier No.:L1082
Specifications
About this item
For use with zeocin resistant strains and strains harboring plasmids such as pCR®-Blunt, pVgRXR, pZeoSV2 series plasmids.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 0.75% Sodium Chloride, 1.5% agar 25 µg/ml zeocin
Ambient shipping conditions. Refrigerate to store.
Made with 0.75% NaCl and pH adjusted to 7.5.