3B Scientific® Cell Anatomy Models Bundle
Catalog #: 470230-928
Supplier: Avantor
Specifications
- Description:Plant and Animal Cell Models
- Cat. no.:470230-928
Specifications
About this item
Modeled After EM Scans.
- Materials resist breakage
- Buy as a set and save
- 3-year manufacturers warranty
These two cell models show the anatomy of plant and animal cells in very high detail. Not just a basic conceptual model, but constructed from scans using electron microscopy. Organelles are presented in a raised relief from the surface, and structures are identified with included keys. Even the removable bases on these models are functional, showing the cells placement with adjacent cells and even showing the plasmodesmata between the plant cells.
Customers Who Bought This Also Bought