SpectraMax® DuoLuc™ Reporter Assay Kit, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
Certificates
About this item
The SpectraMax® DuoLuc™ Reporter Assay Kit offers sensitive detection of dual luciferase reporter gene assays, which are important tools for monitoring gene expression and other cellular events.
- The SpectraMax® DuoLuc™ Reporter Assay Kit enables highly sensitive quantitation of both firefly and Renilla luciferases in mammalian cells at a great price.
- Flash-type luminescence with excellent sensitivity
- Expression measurement in as few as ten cells per well
- Improved accuracy of results by normalizing reporter expression to a control
- Optimized for SpectraMax® i3x, iD3, or L microplate readers with injectors
- Simplified data acquisition and analysis with preconfigured protocol in SoftMax® Pro Software
- Support for your complete solution of reader, software, and assay
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.