SP Wilmad-LabGlass Indented Spherical Reaction Vessels, Conical Flange, SP Industries
Supplier: Avantor
Certificates
About this item
These borosilicate glass reaction vessels have a spherical body with indentations for improved stirring.
Upper end of neck is tooled to obtain a thickened portion and flat top. Top is finely ground for a leak-proof fit with lids LG-8030 to LG-8050.
Customers Who Bought This Also Bought