To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Source:E. coli
- Size:500 µg
- Endotoxin content:Kinetic LAL ≤1 EUs/µg
- Form:Lyophilized
- Protein synonyms:CXCL10|IP-10|CRG-2
- Protein/peptide name:IP-10
- Purity:Reducing and non-reducing SDS PAGE >95%
- Molecular weight:Monomer, 8.7 kDa (76 aa)
- Cat. no.:10822-234
- Supplier No.:400-02-500UG
Specifications
About this item
US-made bioactive recombinant proteins offer rigid quality testing and low manufacturer pricing.
- Strict quality standards
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells and fibroblasts in response to interferon gamma (IFN-ɣ). IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3. IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus.