Malt Extract Agar Plates, Teknova
Supplier: Teknova
Certificates
About this item
Malt extract agar plates are enriched with 1.3% maltose, 0.3% dextrin, 0.23% glycerol, 0.08% peptone, 1.5% agar
- Ambient shipping conditions
- Refrigerate storage
These agar plates are recommended for the growth of yeasts and molds.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.