dCTP CF® Dye Conjugates, Biotium
Supplier: Biotium
Certificates
About this item
CF® dye-dCTP can be used to synthesize probes for in situ hybridization, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
- Synthesize exceptionally bright and photostable fluorescent DNA probes
- Detect by fluorescence microscopy, flow cytometry, or in gel
- Compatible with fixed cells or tissue sections
- Choice of nine CF® dye colors from green to far-red
- Biotiums CF® dyes have superior brightness, photostability, and are water-soluble
Caution: NOTE: CF™594 dCTP is not compatible with PCR labeling of DNA probes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.