Dulbecco's Phosphate-Buffered Saline (DPBS)
Supplier: Promocell
Certificates
About this item
These Dulbecco's PBS solution are used in tissue culture, cell isolation procedures and molecular biology.
- Ready-to-use buffers
- Storage at 4 to 8 °C
Routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.