MColortest™ Carbonate Hardness Test, MilliporeSigma
Supplier: MERCK, S.A. DE C.V. SV
Certificates
About this item
Quick tests, visual colour comparison
- Results in approximately 5 minute
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
Determines carbonate hardness as a measure of the buffering capacity of water, expressed as acid-binding capacity, Acid capacity to pH 4.3 ("SBV", ANC)
Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.