To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:RNA gel bands
- Sample type:Gel matrix
- Target molecule:RNA
- Description:Zymoclean™ Gel RNA Recovery Kit
- Size:50 Preps
- Description:Zymoclean™ Gel RNA Recovery Kit
- Yield:10 µg RNA (binding capacity), ≥6 µl (elution volume)
- Cat. no.:76020-590
Specifications
About this item
The Zymoclean™ Gel RNA Recovery Kit provides a quick purification method for recovery of RNA fragments from agarose gels.
- Quick (30 minute) recovery of purified RNA fragments from agarose gels
- Fast-Spin column technology allows RNA to be eluted into minimal volumes (≥ 6 µL)
- Recovery ≥ 80% for RNA> 500 nt
- High quality RNA (A260/A280 > 1.8, A260/A230 > 1.8) suitable for all downstream RNA-based manipulations
Compatible with MOPS, TAE, and TBE buffered agarose gels (formaldehyde up to 2.0%).
Sample Source: Single- or double-stranded RNA fragments (≥200 nt) resolved in TAE TBE, and MOPS buffered agarose gels. Compatible with formaldehyde to 2.0% (final conc.)
Size Range: RNA fragments ≥500 nt (≥80% recovery)