To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:TBS Dry Blend, Blocking Buffer
- Size:200 mL/Pack
- Cat. no.:CAPI37545
- Supplier No.:37545
Specifications
About this item
SuperBlock (TBS) Blocking Buffer is an optimised TBS solution containing a protein that provides incredibly fast and effective blocking for ELISA, IHC and Western Blot Analysis.
- Block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
- Biotin-free for use with streptavidi
- Available in multiple package sizes
- Non serum protein solution yields a high signal-to-noise ratio
- Stable at 4 °C for one year
The protein based formulation does not contain any immunoglobulins, albumin or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.