Pierce™ Blocker™ BLOTTO Blocking Buffer, Thermo Scientific
Catalog #: CAPI37530
Supplier: Thermo Scientific
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Blocker™ BLOTTO in TBS, Blocking Buffer
- Size:1 L
- Cat. no.:CAPI37530
- Supplier No.:37530
Specifications
About this item
Blocker™ BLOTTO is a ready-to-use 5% solution of non fat powdered milk in Tris-buffered saline for Western blot, ELISA, IHC and other Detection methods.
- Convenient as can be diluted as needed
- Easy to use, formulated with anti-foaming agent and thimerosal-free preservative
- Flexible may be used for multiple applications
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought