Isopropyl Alcohol, Non-Sterile, Texwipe®
Supplier: Texwipe
Certificates
About this item
Non-sterile IPA (isopropyl alcohol) is filtered for cleanliness
- Submicron filtered
- Stream delivery or course spray options are available
- Each bottle is lot coded with an expiration date for easy record keeping
Ideal for surface cleaning, wipe downs and for use with any Texwipe cleanroom wiper.
Delivery: It comes in two convenient dispensers trigger spray (0.5 L) and bulk container (3.8 L).
Packaging: Packing: Filled into cleaned containers, and double-bagged for containers with trigger spray.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.