Masterflex® L/S Pharmed BPT Tubing, Cole Parmer
Supplier: Avantor
Certificates
About this item
Use this tubing for longer pumping tubing life, and excellent biocompatability.
- Non-toxic, non-hemolytic, with extremely low permeability
- Resists acids, alkalies, oxidizing agents, and animal and vegetable oils
- Ideal for tissue and cell culture work
- Heat sealable, bondable, and formable
- Opaque beige finish
Masterflex® L/S pump tubing is manufactured to extremely close tolerances that match the L/S pump heads, ensuring accurate, repeatable flow and long tubing life.
Applications include use in cell and tissue culture work and pharmaceutical uses. This tubing is also good for light-sensitive samples.
Operating temperture range is –20 to –80 °C. Static temperature range is –51 to 132 °C.
Sterilize by EtO, autoclave, or gamma radiation (up to 2.5 Mrad). Repeated autoclaving will not affect overall life.
Certifications: USP Class VI; European Pharmacopeia (3.2.9); FDA; NSF-51; Reach Compliant, RoHS Compliant.
Related Information
Certificates
Search by Lot Number76049-350_24203126
Conformity declaration
Masterflex Pump Systems
Product Literature
Customers Who Bought This Also Bought