Masterflex® L/S Platinum-Cured Silicone Tubing, Cole Parmer
Supplier: Cole-Parmer
Certificates
About this item
Most commonly used pharmaceutical grade silicone tubing for Masterflex® pumps.
- Slightly greater clarity than peroxide-cured tubing
- Smooth surfaces; lower protein binding levels
- Fewer potential leachables
- Ideal for pharmaceutical and biotechnology use
Masterflex® L/S pump tubing is manufactured to extremely close tolerances that match the L/S pump heads, ensuring accurate, repeatable flow and long tubing life.
Applications include use in pharmaceutical, biotech, peristaltic pumping, and food industries.
Operating temperture range is –40 to –100 °C. Static temperature range is –50 to 230 °C.
Sterilize by EtO, autoclave, or gamma radiation (up to 5.0 Mrad). To autoclave, loosely coil in non-linting cloth or paper at 121 °C- 1 bar for 30 minutes.
Certifications: USP Class VI; European Pharmacopeia (3.2.9); FDA; Exceeds 3A Sanitary; Reach Compliant, RoHS Compliant, ADCF Compliant.
Related Information
Certificates
Search by Lot Number76049-298_H033J91019 76049-300_H033178005 76049-300_H033J4R005 76049-300_H033J4U010 76049-300_H033J4U011
Conformity declaration
More
Masterflex Pump Systems
Product Literature
TDS Masterflex® Flow Controller.pdf
Datasheet
Customers Who Bought This Also Bought