Specifications
- Description:2xYT Broth
- Size:5 kgá
- Cat. no.:CA80017-506
- Supplier No.:71755-4
Specifications
About this item
Media is granulated.
- Granulation preserves homogeneity
- Easy to weigh out
- Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out
2xYT Broth Contents: Tryptone 16 g/L; Yeast extract 10 g/L; NaCl 5 g/L (pH 6.8-7.2, at 25 °C).
Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.