Sterilin™ Rnase/Dnase-Free Universal Container, Thermo Scientific
Supplier: Samco
Certificates
About this item
Quality assured containers, ideal for the exacting requirements of molecular biology and genomic research applications.
- Free from RNase, DNase, human DNA, and Pyrogens
- 1/2 turn Quickstart cap for ease of handling
- Excellent leak free performance; tested in accordance with BS EN 14254 Annexe D
- 95kPa compliant at ambient temperature
- Convenient self-standing conical base
- Lot number on each container for traceability
- Aseptically manufactured using controlled, automated cleanroom processes
- Maximum RCF: 9500 xg
Manufactured under controlled cleanroom conditions, this clear robust polypropylene container has a wealth of features which make it a laboratory essential. With a working volume of 25 mL and convenient self-standing conical base, it’s perfect for a wide range of general laboratory uses from sample handling to centrifugation applications.
Caution: Note: For research use only. Not for diagnostic applications.
Customers Who Bought This Also Bought