ContactUsLinkComponent
EZwaste® UN/DOT 20 L Closed System for HPLC Solvent Waste, HDPE Disposable Container, Foxx Life Sciences
EZwaste® UN/DOT 20 L Closed System for HPLC Solvent Waste, HDPE Disposable Container, Foxx Life Sciences
Supplier:  VWR International
Certificates
EZwaste® UN/DOT 20 L Closed System for HPLC Solvent Waste, HDPE Disposable Container, Foxx Life Sciences
EZwaste® UN/DOT 20 L Closed System for HPLC Solvent Waste, HDPE Disposable Container, Foxx Life Sciences
Supplier:  VWR International
Certificates
Specifications
  • Catalog No:
  • 75805-450
  • 75805-448
  • 75805-454
  • 75805-452
  • 75805-456
  • 76018-594
  • 76018-596
  • 76018-598
  • 76018-754
  • 75805-446
  • 76018-758
  • 76018-760
  • 76018-762
  • Filter dimensions:
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Height:
  • 406 mm (15.98")
  • 406 mm (15.98")
  • 406 mm (15.98")
  • 406 mm (15.98")
  • 406 mm (15.98")
  • 406 mm (15.98")