EZFlow® Syringe Filter, CA, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
CA syringe filters are low protein binding, have good flow rates, and are great for use with media and buffers.
- Low protein binding
- Good flow rates
- Great for use with medica, buffers
EZFlow® 13, 25, and 33 mm syringe filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2 and 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet. All materials comply with USP Class VI Standards and are available for both sterile and non-sterile applications.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought