HIMELT Crucibles, Morgan Advanced Materials
Catalog #: CA11019-828
Supplier: MORGANITE ADVANCED MATERIALS CA
Specifications
- Description:HIMELT Crucible, Fused Lip
- Cat. no.:CA11019-828
- Supplier No.:8070000002146
Specifications
About this item
HIMELT high-quality, carbon-bonded silicon carbide crucibles are manufactured using the latest roller-forming techniques and are designed to cater to a range of non-ferrous melting applications.
- Fast melting speed through high consistent thermal conductivity
- Excellent thermal shock resistance
- High resistance to oxidation
- Good erosion resistance
- Good resistance to corrosive attack by chemical treatment agents
HIMELT crucibles are colored bright red. They are available in a comprehensive range of shapes and sizes to suit most end user requirements.
Certifications: Manufactured from premium grade raw materials under an ISO 9001:2008 quality management system.
Customers Who Bought This Also Bought