STARRBIDE Crucibles, Morgan Advanced Materials
Supplier: MORGANITE ADVANCED MATERIALS CA
Certificates
About this item
STARRBIDE is a high-quality, tar-bonded silicon carbide material designed to provide reliable performance across the full range of non-ferrous melting and holding applications.
- Good thermal shock resistance
- Good resistance to attack by chemical treatment agents
- High mechanical strength
STARRBIDE crucibles are colored black. They are available in a comprehensive range of shapes and sizes to suit most end-user requirements.
Certifications: Manufactured to ISO9001:2008 quality standards.
Customers Who Bought This Also Bought