SHOWA NSK24 Nitrile Supported, Chemical Resistant, 14 Glove, Showa
Catalog #: 80090-034
Supplier: Showa
Product available on GSA Advantage®
Specifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Blue
- Glove size:L
- Length:35.6 cm (14")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Cuff style:Gauntlet
- Liner material:Cotton/polyester
- Finish:Rough
- Cat. no.:80090-034
- Supplier No.:NSK24-10
Specifications
About this item
Double dipped Nitrile coating and rough finish provide excellent grip, and protection from chemicals.
- Rough grip
- 14" length
- Highly flexible coating
These gloves are used to handle a wide array of solvents, oils, and acids. Double dipped coating ensures added protection when working with hazardous chemicals.
Customers Who Bought This Also Bought