SHOWA N8 30 mil, Unsupported Neoprene, Chemical Resistant, 18 Glove, Showa
Supplier: Showa
Certificates
About this item
Neoprene is the perfect choice for use with a wide range of chemicals and acids.
- 30 mil thickness
- 18" length
- Neoprene offers a wide range of protection
Gloves are designed to handle chemicals and acids. Latex-free and powder-free.
Specifications
- Catalog No:
- 47742-400
- 47742-402
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Finish:
- Textured
- Textured
- Cuff style:
- Gauntlet
- Gauntlet