SP Wilmad-LabGlass Washing Bottles with Fritted Cylinders, SP Industries
Supplier: Avantor
Certificates
About this item
These gas washing bottles are made of borosilicate glass and have a stopper component that contains gas inlet and outlet tubes.
The lower inlet tube, extending nearly to the base of the bottle, contains a Coarse or Extra-Coarse sealed-on fritted cylindrical disc (12 mm Ext.Ø×25 mm length), providing a stream of fine bubbles into the bottle.
Bottle and stopper are joined by a Standard Taper 29/42 joint.
Delivery: Complete units are supplied with a plastic clamp (LG-1004-110).
Customers Who Bought This Also Bought